Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP1AR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557436
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557436 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557436 Supplier Invitrogen™ Supplier No. PA557436
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GNCCWTQCFG LLRKEAGRLQ RVGGGGGSKY FRTCSRGEHL TIEFENLVES DEGESPGSSH RPLTEEEIVD LRERHYDSIA E Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 94%.

AP1AR is necessary for AP-1 dependent transport between the trans-Golgi network and endosomes. It regulates the membrane association of AP1G1/Gamma1-adaptin, one of the subunits of the AP-1 adapter complex. The direct interaction with AP1G1/Gamma1-adaptin attenuates the release of the AP-1 complex from membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP1AR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AP1AR
Gene Accession No. Q63HQ0
Gene Alias 2c18; AA407621; adapter-related protein complex 1-associated regulatory protein; adaptor related protein complex 1 associated regulatory protein; adaptor-related protein complex 1 associated regulatory protein; adaptor-related protein complex 1-associated regulatory protein; AP-1 complex-associated regulatory protein; AP-1 complex-associated regulatory protein-like; AP1AR; C4orf16; C78909; Gadkin; gamma-1-adaptin brefeldin A resistance protein; gamma1-adaptin brefeldin A resistance protein, gamma-BAR; Gamma-A1-adaptin and kinesin interactor; gamma-BAR; GBAR; PRO0971; RGD1311435
Gene Symbols AP1AR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AP1AR. Recombinant protein control fragment (Product #RP-96658).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55435
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.