Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AP1AR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566133
Description
Immunogen sequence: RIVQQYHPSNN GEYQSSGPED DFESCLRNMK SQYEVFRSSR LSSDATVLTP NTESSCDLMT KTKSTSGNDD STSLDLEWED Highest antigen sequence identity to the following orthologs - mouse 88%, rat 88%.
AP1AR is necessary for AP-1 dependent transport between the trans-Golgi network and endosomes. It regulates the membrane association of AP1G1/Gamma1-adaptin, one of the subunits of the AP-1 adapter complex. The direct interaction with AP1G1/Gamma1-adaptin attenuates the release of the AP-1 complex from membranes.
Specifications
AP1AR | |
Polyclonal | |
Unconjugated | |
AP1AR | |
2c18; AA407621; adapter-related protein complex 1-associated regulatory protein; adaptor related protein complex 1 associated regulatory protein; adaptor-related protein complex 1 associated regulatory protein; adaptor-related protein complex 1-associated regulatory protein; AP-1 complex-associated regulatory protein; AP-1 complex-associated regulatory protein-like; AP1AR; C4orf16; C78909; Gadkin; gamma-1-adaptin brefeldin A resistance protein; gamma1-adaptin brefeldin A resistance protein, gamma-BAR; Gamma-A1-adaptin and kinesin interactor; gamma-BAR; GBAR; PRO0971; RGD1311435 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55435 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q63HQ0 | |
AP1AR | |
Recombinant Human AP1AR. Recombinant protein control fragment (Product #RP-106817). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction