Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP1AR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546203
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546203 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546203 Supplier Invitrogen™ Supplier No. PA546203
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DSTSLDLEWE DEEGMNRMLP MRERSKTEED ILRAALKYSN KKTGSNPTSA Sequence homology: Cow: 93%; Dog: 93%; Horse: 83%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 86%; Rat: 77%.

AP1AR is necessary for AP-1 dependent transport between the trans-Golgi network and endosomes. It regulates the membrane association of AP1G1/Gamma1-adaptin, one of the subunits of the AP-1 adapter complex. The direct interaction with AP1G1/Gamma1-adaptin attenuates the release of the AP-1 complex from membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP1AR
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene AP1AR
Gene Accession No. Q63HQ0
Gene Alias 2c18; AA407621; adapter-related protein complex 1-associated regulatory protein; adaptor related protein complex 1 associated regulatory protein; adaptor-related protein complex 1 associated regulatory protein; adaptor-related protein complex 1-associated regulatory protein; AP-1 complex-associated regulatory protein; AP-1 complex-associated regulatory protein-like; AP1AR; C4orf16; C78909; Gadkin; gamma-1-adaptin brefeldin A resistance protein; gamma1-adaptin brefeldin A resistance protein, gamma-BAR; Gamma-A1-adaptin and kinesin interactor; gamma-BAR; GBAR; PRO0971; RGD1311435
Gene Symbols AP1AR
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human AP1AR (aa 202-251).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55435
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.