Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP1M1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598843
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598843 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598843 Supplier Thermo Scientific Supplier No. PA598843
Add to Cart
Add to Cart

AP1M1 Polyclonal Antibody

AP1M1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen AP1M1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AP1M1
Gene Accession No. Q9BXS5
Gene Alias [m]1A; AA408894; adapter-related protein complex 1 subunit mu-1; adaptor protein complex AP-1 mu-1 subunit; adaptor protein complex AP-1 subunit mu-1; adaptor related protein complex 1 mu 1 subunit; adaptor-related protein complex 1 mu 1 subunit; adaptor-related protein complex 1 mu-1 subunit; adaptor-related protein complex 1 subunit mu-1; adaptor-related protein complex 1, mu 1 subunit; adaptor-related protein complex AP-1, mu subunit 1; Adtm1A; AP-1 complex subunit mu-1; Ap1m1; ap1m1 {ECO:0000312; AP47; AP-mu chain family member mu1A; CLAPM2; clathrin adaptor protein AP47; clathrin assembly protein complex 1 medium chain 1; clathrin assembly protein complex 1 mu-1 medium chain 1; clathrin assembly protein complex 1, medium chain; clathrin assembly protein complex AP1, mu subunit; clathrin coat assembly protein AP47; Clathrin coat-associated protein AP47; Cltnm; golgi adaptor AP-1 47 kDa protein; golgi adaptor HA1/AP1 adaptin mu-1 subunit; HA1 47 kDa subunit; mu1A; MU-1A; mu1A-adaptin; Mu-adaptin 1; RGD:1307653}
Gene Symbols AP1M1
Host Species Rabbit
Immunogen Recombinant Human AP-1 complex subunit mu-1 protein (1-160AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8907
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.