Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP1M2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597011
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597011 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597011 Supplier Invitrogen™ Supplier No. PA597011
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IKYKKNEVFI DVIESVNLLV NANGSVLLSE IVGTIKLKVF LSGMPELRLG LNDRVLFELT GRSKNKSVEL EDVKFHQCVR LSRFDNDRTI SFIPPDGDFE LMSYRLSTQV KPLIWIESVI EKFSHSRVEI MVKAKGQFKK QSVANGVEIS VPVPSDADSP RFKTSVGSAK YVPERNVVIW SIKSFPGGKE YLMRAHFGLP SVEKEEVEGR PPIGVKFEIP YFTVSGIQVR YMKIIEKSGY QALPWVRYIT QSGDYQLRTS; Positive Samples: 22Rv1, BT-474, HepG2, Mouse kidney, Mouse lung, Mouse small intestine; Cellular Location: Cytoplasmic side, Cytoplasmic vesicle, Golgi apparatus, Peripheral membrane protein, clathrin-coated vesicle membrane.

This gene encodes a subunit of the heterotetrameric adaptor-related protein comlex 1 (AP-1), which belongs to the adaptor complexes medium subunits family. This protein is capable of interacting with tyrosine-based sorting signals. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP1M2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AP1M2
Gene Accession No. Q9WVP1, Q9Y6Q5
Gene Alias [m]1B; adaptor protein complex AP-1 mu-2 subunit; adaptor protein complex AP-1 subunit mu-2; adaptor protein complex AP-1, mu 2 subunit; adaptor related protein complex 1 mu 2 subunit; adaptor-related protein complex 1 mu 2 subunit; adaptor-related protein complex 1 mu-2 subunit; adaptor-related protein complex 1 subunit mu-2; adaptor-related protein complex 1, mu 2 subunit; adaptor-related protein complex AP-1, mu 2 subunit; AP-1 complex subunit mu-2; Ap1m2; AP1-mu2; AP-mu chain family member mu1B; clathrin assembly protein complex 1 medium chain 2; clathrin assembly protein complex 1 mu-2 medium chain 2; clathrin coat assembly protein AP47 2; clathrin coat associated protein AP47 2; clathrin-associated adaptor medium chain mu2; D9Ertd818e; golgi adaptor AP-1 47 kDa protein; golgi adaptor HA1/AP1 adaptin mu-2 subunit; HA1 47 kDa subunit 2; HSMU1B; MU1B; MU-1B; Mu1B-adaptin; mu2; Mu-adaptin 2; RGD1561490
Gene Symbols AP1M2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 164-423 of human AP1M2 (NP_0054892).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10053, 11768
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.