Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP1S1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570725
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570725 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570725 Supplier Invitrogen™ Supplier No. PA570725
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

AP1S1 is part of the clathrin coat assembly complex which links clathrin to receptors in coated vesicles. These vesicles are involved in endocytosis and Golgi processing. This protein, as well as beta-prime-adaptin, gamma-adaptin, and the medium (mu) chain AP47, form the AP-1 assembly protein complex located at the Golgi vesicle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP1S1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Ap1s1
Gene Accession No. P61967
Gene Alias adapter-related protein complex 1 sigma-1A subunit; adapter-related protein complex 1 subunit sigma-1A; adaptor protein complex AP-1 sigma-1A subunit; adaptor protein complex AP-1 subunit sigma-1A; adaptor protein complex AP-1, sigma 1; adaptor related protein complex 1 sigma 1 subunit; adaptor related protein complex 1 subunit sigma 1; adaptor-related protein complex 1 sigma 1 subunit; adaptor-related protein complex 1 subunit sigma-1A; adaptor-related protein complex 1, sigma 1 subunit; adaptor-related protein complex AP-1, sigma 1; AP-1 complex subunit sigma-1A; AP19; AP1S1; ap1s1 protein; CLAPS1; Clathrin assembly protein complex 1 sigma-1A small chain; clathrin coat assembly protein AP19; clathrin-associated/assembly/adaptor protein, small 1 (19kD); EKV3; golgi adaptor HA1/AP1 adaptin sigma-1A subunit; HA1 19 kDa subunit; MEDNIK; N-terminus-conjugating E2; sigma 1a subunit of AP-1 clathrin; SIGMA1A; sigma1A subunit of AP-1 clathrin adaptor complex; sigma1A-adaptin; Sigma-adaptin 1A; Ube2w; ubiquitin carrier protein W; ubiquitin-conjugating enzyme E2 W; ubiquitin-protein ligase W; WUGSC:H_DJ0747G18.2; zgc:56704; zgc:85689
Gene Symbols Ap1s1
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of mouse AP1S1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11769
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.