Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP1S1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501570319
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-0319 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0319 Supplier Bethyl Laboratories, Inc Supplier No. A305396AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine

AP1S1 is part of the clathrin coat assembly complex which links clathrin to receptors in coated vesicles. These vesicles are involved in endocytosis and Golgi processing. This protein, as well as beta-prime-adaptin, gamma-adaptin, and the medium (mu) chain AP47, form the AP-1 assembly protein complex located at the Golgi vesicle.

Specifications

Antigen AP1S1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene AP1S1
Gene Accession No. P61966, P61967
Gene Alias AP19, CLAPS1, SIGMA1A, WUGSC:H_DJ0747G18.2, AP-1 complex subunit sigma-1A, HA1 19 kDa subunit, adapter-related protein complex 1 sigma-1A subunit, adaptor protein complex AP-1 sigma-1A subunit, clathrin assembly protein complex 1 sigma-1A small chain, clathrin coat assembly protein AP19, clathrin-associa
Gene Symbols AP1S1
Host Species Rabbit
Immunogen Between 108 and 158
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1174, 11769
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.