Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP1S2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP133101

Catalog No. NBP133101


Only null left
Add to Cart

Description

Description

AP1S2 Polyclonal specifically detects AP1S2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

AP1S2
Polyclonal
Unconjugated
0.1M Tris-Glycine (pH7), 10% Glycerol with 0.01% Thimerosal
AP1S2
Full length human AP1S2 Recombinant protein.
Affinity Purified
RUO
Primary
Not yet tested in any other species.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot, Immunocytochemistry, Immunofluorescence
0.53mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
adaptor-related protein complex 1, sigma 2 subunit, MGC:1902, SIGMA1B, X-linked 59
Rabbit
19 kDa
0.1 mL
Signal Transduction
8905
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only