Learn More
Invitrogen™ AP1S2 Monoclonal Antibody (3B9-G5)

Description
Peptide Sequence: MQFMLLFSRQ GKLRLQKWYV PLSDKEKKKI TRELVQTVLA RKPKMCSFLE WRDLKIVYKR YASLYFCCAI EDQDNELITL EIIHRYVELL DKYFGSVCEL DIIFNFEKAY FILDEFLLGG EVQETSKKNV LKAIEQADLL QEEAETPRSV LEEIGLT.
Specifications
Specifications
| Antigen | AP1S2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 3B9-G5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | AP1S2 |
| Gene Accession No. | P56377 |
| Gene Alias | adapter-related protein complex 1 sigma-1B subunit; adaptor protein complex AP-1 sigma-1B subunit; Adaptor protein complex AP-1 subunit sigma-1B; adaptor related protein complex 1 sigma 2 subunit; adaptor-related protein complex 1 sigma 2 subunit; adaptor-related protein complex 1 subunit sigma-1B; adaptor-related protein complex 1, sigma 2 subunit; AP-1 complex subunit sigma-2; AP1S2; clathrin adaptor complex AP1 sigma 1B subunit; clathrin assembly protein complex 1 sigma-1B small chain; DC22; golgi adaptor HA1/AP1 adaptin sigma-1B subunit; MGC:1902; MRX59; MRXS21; MRXS5; MRXSF; PGS; Sigma 1B subunit of AP-1 clathrin; SIGMA1B; sigma1B-adaptin; Sigma-adaptin 1B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.