Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP1S2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562081 Supplier Invitrogen™ Supplier No. PA562081
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QADLLQEKTE TMYHSKSFIG FKKAY Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 44%.

Adaptor protein complex 1 is found at the cytoplasmic face of coated vesicles located at the Golgi complex, where it mediates both the recruitment of clathrin to the membrane and the recognition of sorting signals within the cytosolic tails of transmembrane receptors. This complex is a heterotetramer composed of two large, one medium, and one small adaptin subunit. The protein encoded by this gene serves as the small subunit of this complex and is a member of the adaptin protein family. Transcript variants utilizing alternative polyadenylation signals exist for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP1S2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AP1S2
Gene Accession No. P56377
Gene Alias adapter-related protein complex 1 sigma-1B subunit; adaptor protein complex AP-1 sigma-1B subunit; Adaptor protein complex AP-1 subunit sigma-1B; adaptor related protein complex 1 sigma 2 subunit; adaptor-related protein complex 1 sigma 2 subunit; adaptor-related protein complex 1 subunit sigma-1B; adaptor-related protein complex 1, sigma 2 subunit; AP-1 complex subunit sigma-2; AP1S2; clathrin adaptor complex AP1 sigma 1B subunit; clathrin assembly protein complex 1 sigma-1B small chain; DC22; golgi adaptor HA1/AP1 adaptin sigma-1B subunit; MGC:1902; MRX59; MRXS21; MRXS5; MRXSF; PGS; Sigma 1B subunit of AP-1 clathrin; SIGMA1B; sigma1B-adaptin; Sigma-adaptin 1B
Gene Symbols AP1S2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AP1S2. Recombinant protein control fragment (Product #RP-100849).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8905
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.