Learn More
Invitrogen™ AP1S2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA522018
Description
Recommended positive controls: A431, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (92%), Xenopus laevis (96%), Chicken (99%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Adaptor protein complex 1 is found at the cytoplasmic face of coated vesicles located at the Golgi complex, where it mediates both the recruitment of clathrin to the membrane and the recognition of sorting signals within the cytosolic tails of transmembrane receptors. This complex is a heterotetramer composed of two large, one medium, and one small adaptin subunit. The protein encoded by this gene serves as the small subunit of this complex and is a member of the adaptin protein family. Transcript variants utilizing alternative polyadenylation signals exist for this gene.
Specifications
AP1S2 | |
Polyclonal | |
Unconjugated | |
AP1S2 | |
adapter-related protein complex 1 sigma-1B subunit; adaptor protein complex AP-1 sigma-1B subunit; Adaptor protein complex AP-1 subunit sigma-1B; adaptor related protein complex 1 sigma 2 subunit; adaptor-related protein complex 1 sigma 2 subunit; adaptor-related protein complex 1 subunit sigma-1B; adaptor-related protein complex 1, sigma 2 subunit; AP-1 complex subunit sigma-2; AP1S2; clathrin adaptor complex AP1 sigma 1B subunit; clathrin assembly protein complex 1 sigma-1B small chain; DC22; golgi adaptor HA1/AP1 adaptin sigma-1B subunit; MGC:1902; MRX59; MRXS21; MRXS5; MRXSF; PGS; Sigma 1B subunit of AP-1 clathrin; SIGMA1B; sigma1B-adaptin; Sigma-adaptin 1B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8905 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.53 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P56377 | |
AP1S2 | |
Full length human AP1S2 Recombinant protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.