Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP1S3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597465
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597465 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597465 Supplier Invitrogen™ Supplier No. PA597465
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AP1S3 Polyclonal Antibody for Western Blot, ELISA

Subunit of clathrin-associated adaptor protein complex 1 that plays a role in protein sorting in the late-Golgi/trans-Golgi network (TGN) and/or endosomes. The AP complexes mediate both the recruitment of clathrin to membranes and the recognition of sorting signals within the cytosolic tails of transmembrane cargo molecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP1S3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene AP1S3
Gene Accession No. Q96PC3
Gene Alias [s]3A; 1190009B22; adapter-related protein complex 1 sigma-1C subunit; adapter-related protein complex 1 subunit sigma-1C; adaptor protein complex AP-1 sigma-1C subunit; adaptor protein complex AP-1 subunit sigma-1C; adaptor related protein complex 1 sigma 3 subunit; adaptor-related protein complex 1 sigma 3 subunit; adaptor-related protein complex 1 subunit sigma-1C; adaptor-related protein complex 1, sigma 3 subunit; adaptor-related protein complex AP-1, sigma 3; AP-1 complex subunit sigma-3; Ap1s3; Clathrin assembly protein complex 1 sigma-1C small chain; golgi adaptor HA1/AP1 adaptin sigma-1C subunit; Jr2; PSORS15; sigma 1C; sigma 1C adaptin; sigma 1C subunit of AP-1 clathrin; sigma1C-adaptin; Sigma-adaptin 1C
Gene Symbols AP1S3
Host Species Rabbit
Immunogen Recombinant Human AP-1 complex subunit sigma-3 protein (1-104AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130340
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.