Learn More
Invitrogen™ AP1S3 Polyclonal Antibody

Description
AP1S3 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | AP1S3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | AP1S3 |
| Gene Accession No. | Q7TN05, Q96PC3 |
| Gene Alias | [s]3A; 1190009B22; adapter-related protein complex 1 sigma-1C subunit; adapter-related protein complex 1 subunit sigma-1C; adaptor protein complex AP-1 sigma-1C subunit; adaptor protein complex AP-1 subunit sigma-1C; adaptor related protein complex 1 sigma 3 subunit; adaptor-related protein complex 1 sigma 3 subunit; adaptor-related protein complex 1 subunit sigma-1C; adaptor-related protein complex 1, sigma 3 subunit; adaptor-related protein complex AP-1, sigma 3; AP-1 complex subunit sigma-3; Ap1s3; Clathrin assembly protein complex 1 sigma-1C small chain; golgi adaptor HA1/AP1 adaptin sigma-1C subunit; Jr2; PSORS15; sigma 1C; sigma 1C adaptin; sigma 1C subunit of AP-1 clathrin; sigma1C-adaptin; Sigma-adaptin 1C |
| Gene Symbols | AP1S3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.