Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP2 alpha + beta Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501982271
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2271 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2271 Supplier Bioss Supplier No. BS12480R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a transcription factor that binds the consensus sequence 5′-GCCNNNGGC-3′. The encoded protein functions as either a homodimer or as a heterodimer with similar family members. This protein activates the transcription of some genes while inhibiting the transcription of others. Defects in this gene are a cause of branchiooculofacial syndrome. Three transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen AP2 alpha + beta
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TFAP2A
Gene Accession No. P05549, P34056, P58197, Q61313, Q92481
Gene Alias activating enhancer binding protein 2 beta; Activating enhancer-binding protein 2-alpha; Activating enhancer-binding protein 2-beta; Activator protein 2; AP-2; AP-2 transcription factor; AP-2alpha; AP2-alpha; AP-2B; AP2-B; AP2-beta; AP2TF; BOFS; FLJ51761; TFAP2; TFAP2A; TFAP2B; transcription factor AP-2 alpha; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); transcription factor AP-2 beta; transcription factor AP-2 beta (activating enhancer binding protein 2 beta); Transcription factor AP-2-alpha; Transcription factor AP-2-beta
Gene Symbols TFAP2A, TFAP2B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human AP2 alpha + beta.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21418, 21419, 301285, 306862, 7020, 7021
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.