Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP2 alpha Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA131776
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA131776 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA131776 Supplier Invitrogen™ Supplier No. PA131776
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TFAP2A is a sequence-specific DNA-binding protein that interacts with inducible viral and cellular enhancer elements to regulate transcription of selected genes. AP-2 factors bind to the consensus sequence 5'-GCCNNNGGC-3' and activate genes involved in a large spectrum of important biological functions including proper eye, face, body wall, limbs and neural tube development. They also suppress a number of genes including MCAM/MUC18, C/EBP alpha and MYC. TFAP2A is the only AP-2 protein required for early morphogenesis of the lens vesicle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP2 alpha
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene TFAP2A
Gene Accession No. P05549
Gene Alias activating enhancer-binding protein 2 alpha; activating enhancer-binding protein 2-alpha; Activator protein 2; activator protein 2 alpha variant 4; activator protein 2 alpha variant 6; activator protein 2 alpha variant 7; AP2; AP-2; Ap-2 (a); AP-2 transcription factor; AP2alpha; AP-2alpha; AP2-alpha; Ap2tf; BOFS; ERF-1; FLJ51761; OTTHUMP00000214240; OTTHUMP00000214243; Tcfap2a; TFAP2; TFAP2A; transcription factor AP2; transcription factor AP-2 alpha; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); transcription factor AP-2, alpha; transcription factor AP-2-alpha; transcription factor AP-2-alpha-like; transcription factor; AP-2
Gene Symbols TFAP2A
Host Species Goat
Immunogen Synthetic peptide corresponding to the N-Terminal region near aa 1-25 of Human AP2A1 and AP2A2 proteins.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7020
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.