Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP2 alpha/beta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599259
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599259 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599259 Supplier Invitrogen™ Supplier No. PA599259
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AP2 alpha/beta.

The protein encoded by this gene is a transcription factor that binds the consensus sequence 5'-GCCNNNGGC-3'. The encoded protein functions as either a homodimer or as a heterodimer with similar family members. This protein activates the transcription of some genes while inhibiting the transcription of others. Defects in this gene are a cause of branchiooculofacial syndrome. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP2 alpha/beta
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TFAP2A
Gene Accession No. P05549, P34056, P58197, Q61313, Q92481
Gene Alias activating enhancer binding protein 2 beta; activating enhancer-binding protein 2 alpha; activating enhancer-binding protein 2 beta; activating enhancer-binding protein 2-alpha; Activating enhancer-binding protein 2-beta; activator protein 2; AI606113; Ap2; AP-2; Ap-2 (a); AP-2 transcription factor; AP-2(beta); AP2alpha; AP-2alpha; AP2-alpha; AP-2B; AP2-B; AP2-beta; AP2TF; BOFS; E130018K07Rik; FLJ51761; Tcfap2a; Tcfap2b; TFAP2; Tfap2a; TFAP2B; transcription factor AP-2 alpha; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); transcription factor AP-2 beta; transcription factor AP-2 beta (activating enhancer binding protein 2 beta); transcription factor AP-2, alpha; transcription factor AP-2-alpha; Transcription factor AP-2-beta
Gene Symbols TFAP2A, TFAP2B
Host Species Rabbit
Immunogen A synthesized peptide derived from human TFAP2A(Accession P05549), corresponding to amino acid residues K411-K437.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21418, 21419, 301285, 306862, 7020, 7021
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.