Learn More
Invitrogen™ AP2 alpha Recombinant Rabbit Monoclonal Antibody (24GB2485)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA557769

Description
AP2 alpha Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
TFAP2A is a sequence-specific DNA-binding protein that interacts with inducible viral and cellular enhancer elements to regulate transcription of selected genes. AP-2 factors bind to the consensus sequence 5'-GCCNNNGGC-3' and activate genes involved in a large spectrum of important biological functions including proper eye, face, body wall, limbs and neural tube development. They also suppress a number of genes including MCAM/MUC18, C/EBP alpha and MYC. TFAP2A is the only AP-2 protein required for early morphogenesis of the lens vesicle.
Specifications
AP2 alpha | |
Recombinant Monoclonal | |
Unconjugated | |
TFAP2A | |
activating enhancer-binding protein 2 alpha; activating enhancer-binding protein 2-alpha; Activator protein 2; activator protein 2 alpha variant 4; activator protein 2 alpha variant 6; activator protein 2 alpha variant 7; AP2; AP-2; Ap-2 (a); AP-2 transcription factor; AP2alpha; AP-2alpha; AP2-alpha; Ap2tf; BOFS; ERF-1; FLJ51761; OTTHUMP00000214240; OTTHUMP00000214243; Tcfap2a; TFAP2; TFAP2A; transcription factor AP2; transcription factor AP-2 alpha; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); transcription factor AP-2, alpha; transcription factor AP-2-alpha; transcription factor AP-2-alpha-like; transcription factor; AP-2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
306862, 7020 | |
-20°C | |
Liquid |
Flow Cytometry, Western Blot, Immunocytochemistry | |
24GB2485 | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P05549, P58197 | |
TFAP2A | |
A synthesized peptide derived from human AP2 alpha (aa 100-140). | |
20 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.