Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP2 beta Monoclonal Antibody (6E7H11)
GREENER_CHOICE

Catalog No. PIMA531926
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531926 Supplier Invitrogen™ Supplier No. MA531926
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31926 has been tested in indirect ELISA.

This gene encodes a member of the AP-2 family of transcription factors. AP-2 proteins form homo- or hetero-dimers with other AP-2 family members and bind specific DNA sequences. They are thought to stimulate cell proliferation and suppress terminal differentiation of specific cell types during embryonic development. Specific AP-2 family members differ in their expression patterns and binding affinity for different promoters. This protein functions as both a transcriptional activator and repressor. Mutations in this gene result in autosomal dominant Char syndrome, suggesting that this gene functions in the differentiation of neural crest cell derivatives.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP2 beta
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6E7H11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Tfap2b
Gene Accession No. Q92481
Gene Alias activating enhancer binding protein 2 beta; activating enhancer-binding protein 2 beta; activating enhancer-binding protein 2-beta; AI606113; AP-2(beta); AP-2B; AP2-B; AP2-beta; E130018K07Rik; Tcfap2b; Tfap2b; transcription factor AP-2 beta; transcription factor AP-2 beta (activating enhancer binding protein 2 beta); transcription factor AP-2-beta
Gene Symbols Tfap2b
Host Species Mouse
Immunogen Purified recombinant fragment of human TFAP2B (AA: 84-193) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7021
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.