Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP2 gamma Monoclonal Antibody (AP2g 6E4/4)
GREENER_CHOICE

Catalog No. MA191374
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA191374 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA191374 Supplier Invitrogen™ Supplier No. MA191374
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-91374 detects AP2 gamma from mouse, human samples.

The protein encoded by this gene is a sequence-specific DNA-binding transcription factor involved in the activation of several developmental genes. The encoded protein can act as either a homodimer or heterodimer with other family members and is induced during retinoic acid-mediated differentiation. It plays a role in the development of the eyes, face, body wall, limbs, and neural tube.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP2 gamma
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone AP2g 6E4/4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TFAP2C
Gene Accession No. Q61312, Q92754
Gene Alias AA409384; activating enhancer binding protein 2 gamma; activating enhancer-binding protein 2 gamma; activator protein 2 gamma; AP-2.2; AP2gamma; AP2-GAMMA; ERF1; estrogen receptor factor 1; hAP-2g; stimulated by retinoic acid gene 2; Stra2; Tcfap2c; TFAP2C; TFAP2G; Transcription factor AP-2 gamma; transcription factor AP-2 gamma (activating enhancer binding protein 2 gamma); transcription factor AP-2 gamma (activating enhancer-binding protein 2 gamma); transcription factor AP-2, gamma; transcription factor ERF-1
Gene Symbols TFAP2C
Host Species Mouse
Immunogen Bacterial AP-2a/AP-2g fusion protein.
Purification Method IgG Fraction
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21420, 7022
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.