Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP2 gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581066
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581066 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581066 Supplier Invitrogen™ Supplier No. PA581066
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human TFAP2C/AP2-GAMMA.

The protein encoded by this gene is a sequence-specific DNA-binding transcription factor involved in the activation of several developmental genes. The encoded protein can act as either a homodimer or heterodimer with other family members and is induced during retinoic acid-mediated differentiation. It plays a role in the development of the eyes, face, body wall, limbs, and neural tube.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP2 gamma
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TFAP2C
Gene Accession No. Q92754
Gene Alias AA409384; activating enhancer binding protein 2 gamma; activating enhancer-binding protein 2 gamma; activator protein 2 gamma; AP-2.2; AP2gamma; AP2-GAMMA; ERF1; estrogen receptor factor 1; hAP-2g; stimulated by retinoic acid gene 2; Stra2; Tcfap2c; TFAP2C; TFAP2G; Transcription factor AP-2 gamma; transcription factor AP-2 gamma (activating enhancer binding protein 2 gamma); transcription factor AP-2 gamma (activating enhancer-binding protein 2 gamma); transcription factor AP-2, gamma; transcription factor ERF-1
Gene Symbols TFAP2C
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of the Human TFAP2C/AP2-GAMMA.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7022
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.