Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP2 gamma Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987434
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7434 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7434 Supplier Bioss Supplier No. BS6694R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a sequence-specific DNA-binding transcription factor involved in the activation of several developmental genes. The encoded protein can act as either a homodimer or heterodimer with other family members and is induced during retinoic acid-mediated differentiation. It plays a role in the development of the eyes, face, body wall, limbs, and neural tube.

Specifications

Antigen AP2 gamma
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TFAP2C
Gene Accession No. Q92754
Gene Alias AA409384; activating enhancer binding protein 2 gamma; Activating enhancer-binding protein 2 gamma; activator protein 2 gamma; AP-2.2; AP2gamma; AP2-gamma; ERF1; estrogen receptor factor 1; hAP-2g; stimulated by retinoic acid gene 2; Stra2; Tcfap2c; TFAP2C; TFAP2G; Transcription factor AP-2 gamma; transcription factor AP-2 gamma (activating enhancer binding protein 2 gamma); transcription factor AP-2 gamma (activating enhancer-binding protein 2 gamma); transcription factor AP-2, gamma; Transcription factor ERF-1
Gene Symbols TFAP2C
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TFAP2C.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7022
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.