Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AP2A1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142994
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize reported isoform 1 (NP_055018.2) only. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
This gene encodes the alpha 1 adaptin subunit of the adaptor protein 2 (AP-2) complex found in clathrin coated vesicles. The AP-2 complex is a heterotetramer consisting of two large adaptins (alpha or beta), a medium adaptin (mu), and a small adaptin (sigma). The complex is part of the protein coat on the cytoplasmic face of coated vesicles which links clathrin to receptors in vesicles. Alternative splicing of this gene results in two transcript variants encoding two different isoforms. A third transcript variant has been described, but its full length nature has not been determined. [provided by RefSeq, Jul 2008].
Specifications
AP2A1 | |
Polyclonal | |
Unconjugated | |
AP2A1 | |
AP2A1 | |
Peptide with sequence CELEPPAPESPMALLADPAPAAD. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O95782, P17426 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
11771, 160 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction