Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AP2A2 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299239APC
Description
AP2A2 Polyclonal antibody specifically detects AP2A2 in Human samples. It is validated for Western BlotSpecifications
AP2A2 | |
Polyclonal | |
Western Blot | |
100 kDa coated vesicle protein C, Adapter-related protein complex 2 alpha-2 subunit, adaptin, alpha B, Adaptor protein complex AP-2 subunit alpha-2, adaptor-related protein complex 2, alpha 2 subunit, alpha2-adaptin, Alpha-adaptin C, alpha-adaptin C; Huntingtin interacting protein J, AP-2 complex subunit alpha-2, Clathrin assembly protein complex 2 alpha-C large chain, clathrin-associated/assembly/adaptor protein, large, alpha 2, DKFZP564D1864, HIP9ADTAB, Huntingtin yeast partner J, Huntingtin-interacting protein 9, Huntingtin-interacting protein J, HYPJCLAPA2, KIAA0899HIP-9, Plasma membrane adaptor HA2/AP2 adaptin alpha C subunit | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human AP2A2. | |
0.1 mL | |
Neurodegeneration, Neuroscience | |
161 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction