Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP2A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576733
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576733 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576733 Supplier Invitrogen™ Supplier No. PA576733
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Adaptins are components of the adaptor complexes which link clathrin to receptors in coated vesicles. Clathrin-associated protein complexes are believed to interact with the cytoplasmic tails of membrane proteins, leading to their selection and concentration. Alpha adaptin is a subunit of the plasma membrane adaptor. It binds polyphosphoinositides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP2A2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene AP2A2
Gene Accession No. O94973, P17427
Gene Alias 100 kDa coated vesicle protein C; 2410074K14Rik; adapter-related protein complex 2 alpha-2 subunit; adapter-related protein complex 2 subunit alpha-2; adaptin alpha A; Adaptin alpha B; adaptin, alpha B; adaptor protein complex AP-2 subunit alpha-2; adaptor protein complex AP-2, alpha 2 subunit; adaptor related protein complex 2 alpha 2 subunit; adaptor related protein complex 2, alpha 2 subunit; adaptor-related protein complex 2 subunit alpha-2; adaptor-related protein complex 2, alpha 2 subunit; adaptor-related protein complex AP-2, alpha 2 subunit; ADTAA; ADTAB; AF006990; Alpha adaptin A; Alpha adaptin C; Alpha1 adaptin; Alpha2 adaptin; alpha2-adaptin; alpha-adaptin C; alpha-adaptin C; Huntingtin interacting protein J; alpha-C adaptin; AP2 alpha; AP-2 complex subunit alpha-2; AP2A1; Ap2a2; AW146353; C78001; CLAPA1; CLAPA2; Clathrin assembly; Clathrin assembly protein complex 2 alpha-C large chain; clathrin-associated/assembly/adaptor protein, large, alpha 2; HIP 9; HIP9; HIP-9; Huntingtin yeast partner J; huntingtin-interacting protein 9; Huntingtin-interacting protein J; HYPJ; KIAA0899; L25; Plasma membrane adaptor HA2/AP2 adaptin alpha C subunit
Gene Symbols AP2A2
Host Species Rabbit
Immunogen Recombinant full length Human AP2A2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11772, 161
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.