Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AP2A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112102
Description
This antibody reacts with: Human, Mouse, Rat (Species predicted to react based on 100% sequence homology).
Adaptins are components of the adaptor complexes which link clathrin to receptors in coated vesicles. Clathrin-associated protein complexes are believed to interact with the cytoplasmic tails of membrane proteins, leading to their selection and concentration. Alpha adaptin is a subunit of the plasma membrane adaptor. It binds polyphosphoinositides.
Specifications
| AP2A2 | |
| Polyclonal | |
| Unconjugated | |
| AP2A2 | |
| 100 kDa coated vesicle protein C; 2410074K14Rik; adapter-related protein complex 2 alpha-2 subunit; adapter-related protein complex 2 subunit alpha-2; adaptin alpha A; Adaptin alpha B; adaptin, alpha B; adaptor protein complex AP-2 subunit alpha-2; adaptor protein complex AP-2, alpha 2 subunit; adaptor related protein complex 2 alpha 2 subunit; adaptor related protein complex 2, alpha 2 subunit; adaptor-related protein complex 2 subunit alpha-2; adaptor-related protein complex 2, alpha 2 subunit; adaptor-related protein complex AP-2, alpha 2 subunit; ADTAA; ADTAB; AF006990; Alpha adaptin A; Alpha adaptin C; Alpha1 adaptin; Alpha2 adaptin; alpha2-adaptin; alpha-adaptin C; alpha-adaptin C; Huntingtin interacting protein J; alpha-C adaptin; AP2 alpha; AP-2 complex subunit alpha-2; AP2A1; Ap2a2; AW146353; C78001; CLAPA1; CLAPA2; Clathrin assembly; Clathrin assembly protein complex 2 alpha-C large chain; clathrin-associated/assembly/adaptor protein, large, alpha 2; HIP 9; HIP9; HIP-9; Huntingtin yeast partner J; huntingtin-interacting protein 9; Huntingtin-interacting protein J; HYPJ; KIAA0899; L25; Plasma membrane adaptor HA2/AP2 adaptin alpha C subunit | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 161 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| O94973 | |
| AP2A2 | |
| A synthetic peptide corresponding to the C-terminus of the Human Ap2a2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction