Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP2AGoat anti-Canine, Chicken, Chimpanzee, Human, Mouse, Non-human primate, Rat, Polyclonal, Rockland™
SDP

Catalog No. 501991558
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1558 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1558 Supplier Rockland Immunochemicals Supplier No. 600101287
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human AP2A. A BLAST analysis was used to suggest reactivity with AP2A1 and AP2A2 protein from human, chimpanzee, orangutan, mouse, rat, chicken and dog based on 100% homology for the immunogen sequence. Cross reactivity with AP2A proteins from other sources is not known.

Component of the adaptor protein complex 2 (AP-2). Adaptor protein complexes function in protein transport via transport vesicles in different membrane traffic pathways. Adaptor protein complexes are vesicle coat components and appear to be involved in cargo selection and vesicle formation. AP-2 is involved in clathrin-dependent endocytosis in which cargo proteins are incorporated into vesicles surrounded by clathrin (clathrin-coated vesicles, CCVs) which are destined for fusion with the early endosome. The clathrin lattice serves as a mechanical scaffold but is itself unable to bind directly to membrane components. Clathrin-associated adaptor protein (AP) complexes which can bind directly to both the clathrin lattice and to the lipid and protein components of membranes are considered to be the major clathrin adaptors contributing the CCV formation. AP-2 also serves as a cargo receptor to selectively sort the membrane proteins involved in receptor-mediated endocytosis. AP-2 seems to play a role in the recycling of synaptic vesicle membranes from the presynaptic surface. AP-2 recognizes Y-X-X-[FILMV] (Y-X-X-Phi) and [ED]-X-X-X-L-[LI] endocytosis signal motifs within the cytosolic tails of transmembrane cargo molecules. AP-2 may also play a role in maintaining normal post-endocytic trafficking through the ARF6-regulated, non-clathrin pathway. The AP-2 alpha subunit binds polyphosphoinositide-containing lipids, positioning AP-2 on the membrane. The AP-2 alpha subunit acts via its C-terminal appendage domain as a scaffolding platform for endocytic accessory proteins. The AP-2 alpha and AP-2 sigma subunits are thought to contribute to the recognition of the [ED]-X-X-X-L-[LI] motif.

Specifications

Antigen AP2A
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.7 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Ap2a1
Gene Accession No. O95782, P17426
Gene Alias 100 kDa coated vesicle protein A; adapter-related protein complex 2 alpha-1 subunit; adapter-related protein complex 2 subunit alpha-1; adaptin, alpha A; Adaptor protein complex AP-2 subunit alpha-1; adaptor protein complex AP-2, alpha 1 subunit; adaptor related protein complex 2 alpha 1 subunit; adaptor related protein complex 2, alpha 1 subunit; Adaptor-related protein complex 2 subunit alpha-1; adaptor-related protein complex 2, alpha 1 subunit; adaptor-related protein complex AP-2, alpha 1 subunit; ADTAA; ADTAB; Alpha1-adaptin; alpha2-adaptin; Alpha-adaptin A; alpha-adaptin C; AP2; AP-2 complex subunit alpha-1; AP2A1; AP2-ALPHA; BOS_17797; CLAPA1; CLAPA2; Clathrin assembly protein complex 2 alpha-A large chain; clathrin-associated/assembly/adaptor protein, large, alpha 1; HIP9; HIP-9; HYPJ; Plasma membrane adaptor HA2/AP2 adaptin alpha A subunit
Gene Symbols Ap2a1
Host Species Goat
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11771, 160, 308578, 468956, 484375
Target Species Canine, Chicken, Chimpanzee, Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.