Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AP2M1 Antibody (4F1B2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | AP2M1 |
---|---|
Clone | 4F1B2 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61866
![]() |
Novus Biologicals
NBP261866 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
AP2M1 Monoclonal specifically detects AP2M1 in Human, Monkey samples. It is validated for Western Blot, ELISA.Specifications
AP2M1 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Human, Monkey | |
1173 | |
Purified recombinant fragment of human AP2M1 (AA: 298-435) expressed in E. Coli. | |
Primary | |
49.7 kDa |
4F1B2 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
Phospho Specific, Signal Transduction | |
Adaptin-mu2, adaptor-related protein complex 2, mu 1 subunit, AP-2 mu chain, Clathrin assembly protein complex 2 medium chain, clathrin coat adaptor protein AP50, Clathrin coat assembly protein AP50, Clathrin coat-associated protein AP50, clathrin-associated/assembly/adaptor protein, medium 1, HA2 50 kDa subunit, KIAA0109, mu subunit, Plasma membrane adaptor AP-2 50 kDa protein, plasma membrane adaptor AP-2 50kDA protein | |
AP2M1 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title