Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP2S1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102937
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102937 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102937 Supplier Invitrogen™ Supplier No. PA5102937
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AP2S1.

Component of the adaptor protein complex 2 (AP-2). Adaptor protein complexes function in protein Transport via Transport vesicles in different membrane traffic pathways. Adaptor protein complexes are vesicle coat components and appear to be involved in cargo selection and vesicle formation. AP-2 is involved in clathrin-dependent endocytosis in which cargo proteins are incorporated into vesicles surrounded by clathrin (clathrin- coated vesicles, CCVs) which are destined for fusion with the early endosome. The clathrin lattice serves as a mechanical scaffold but is itself unable to bind directly to membrane components. Clathrin-associated adaptor protein (AP) complexes which can bind directly to both the clathrin lattice and to the lipid and protein components of membranes are considered to be the major clathrin adaptors contributing the CCV formation. AP-2 also serves as a cargo receptor to selectively sort the membrane proteins involved in receptor-mediated endocytosis. AP-2 seems to play a role in the recycling of synaptic vesicle membranes from the presynaptic surface. AP-2 recognizes Y-X-X-[FILMV] (Y-X-X-Phi) and [ED]-X-X-X-L-[LI] endocytosis signal motifs within the cytosolic tails of transmembrane cargo molecules. AP-2 may also play a role in maintaining normal post-endocytic trafficking through the ARF6-regulated, non-clathrin pathway. The AP-2 alpha and AP-2 sigma subunits are thought to contribute to the recognition of the [ED]-X-X-X-L-[LI] motif. May also play a role in extracellular calcium homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP2S1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AP2S1
Gene Accession No. P53680
Gene Alias adapter-related protein complex 2 sigma subunit; adapter-related protein complex 2 subunit sigma; adaptor protein complex AP-2 subunit sigma; adaptor related protein complex 2 sigma 1 subunit; adaptor-related protein complex 2 sigma 1 subunit; adaptor-related protein complex 2 subunit sigma; adaptor-related protein complex 2, sigma 1 subunit; AI043088; Ap17; AP17-DELTA; AP-2 complex subunit sigma; AP2S1; Claps2; Clathrin assembly protein 2 sigma small chain; clathrin assembly protein 2 small chain; clathrin coat assembly protein AP17; Clathrin coat-associated protein AP17; clathrin-associated protein 17; clathrin-associated/assembly/adaptor protein, small 2 (17kD); FBH3; FBHOk; HA2 17 kDa subunit; HHC3; Plasma membrane adaptor AP-2 17 kDa protein; Sigma2-adaptin; sigma-adaptin 3b
Gene Symbols AP2S1
Host Species Rabbit
Immunogen A synthesized peptide derived from human AP2S1(Accession P53680), corresponding to amino acid residues M1-V39.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1175
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.