Learn More
Invitrogen™ AP2S1 Recombinant Rabbit Monoclonal Antibody (4G1G1)

Description
Positive test controls include: Mouse brain. Immunogen sequence: MIRFILIQNR AGKTRLAKWY MQFDDDEKQK LIEEVHAVVT VRDAKHTNFV EFRNFKIIYR RYAGLYFCIC VDVNDNNLAY LEAIHNFVEV LNEYFHNVCE.
Specifications
Specifications
| Antigen | AP2S1 |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 4G1G1 |
| Concentration | 1.88 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | AP2S1 |
| Gene Accession No. | P62743 |
| Gene Alias | adapter-related protein complex 2 sigma subunit; adapter-related protein complex 2 subunit sigma; adaptor protein complex AP-2 subunit sigma; adaptor related protein complex 2 sigma 1 subunit; adaptor-related protein complex 2 sigma 1 subunit; adaptor-related protein complex 2 subunit sigma; adaptor-related protein complex 2, sigma 1 subunit; AI043088; Ap17; AP17-DELTA; AP-2 complex subunit sigma; AP2S1; Claps2; Clathrin assembly protein 2 sigma small chain; clathrin assembly protein 2 small chain; clathrin coat assembly protein AP17; Clathrin coat-associated protein AP17; clathrin-associated protein 17; clathrin-associated/assembly/adaptor protein, small 2 (17kD); FBH3; FBHOk; HA2 17 kDa subunit; HHC3; Plasma membrane adaptor AP-2 17 kDa protein; Sigma2-adaptin; sigma-adaptin 3b |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.