Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP3B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121055
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121055 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121055 Supplier Thermo Scientific Supplier No. PA5121055
Add to Cart
Add to Cart
This item is not returnable. View return policy

AP3B1 Polyclonal Antibody

AP3B1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen AP3B1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ap3b1
Gene Accession No. O00203, Q9Z1T1
Gene Alias adapter-related protein complex 3 subunit beta-1; adaptor protein complex AP-3 beta-1 subunit; adaptor protein complex AP-3 subunit beta-1; adaptor related protein complex 3 beta 1 subunit; adaptor related protein complex 3 subunit beta 1; adaptor related protein complex 3, beta 1 subunit; adaptor-related protein complex 3 subunit beta-1; adaptor-related protein complex 3, beta 1 subunit; adaptor-related protein complex AP3 beta 1 subunit; adaptor-related protein complex AP-3, beta 1 subunit; ADTB3; ADTB3A; ap3; AP-3; AP-3 complex beta-3A subunit; AP-3 complex subunit beta-1; AP-3 complex subunit beta-1-like protein; AP3B1; AU015684; beta3A; Beta-3A-adaptin; beta3A-adaptin; C78395; Clathrin assembly protein complex 3 beta-1 large chain; HPS; Hps2; PE; pearl; recombination induced mutation 2; rim2; Unknown (protein for MGC:128992)
Gene Symbols Ap3b1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 895-1094 of human AP3B1 (NP_003655.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11774, 309969, 8546
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.