Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AP3B1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529858
Description
Recommended positive controls: HeLa, HepG2. Predicted reactivity: Mouse (98%), Xenopus laevis (96%), Dog (100%), Pig (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
AP3B1 is a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. Structurally, AP3B1 is part of the heterotetrameric AP-3 protein complex which interacts with the scaffolding protein clathrin. Mutations in AP3B1 are associated with Hermansky-Pudlak syndrome type 2.
Specifications
AP3B1 | |
Polyclonal | |
Unconjugated | |
Ap3b1 | |
adapter-related protein complex 3 subunit beta-1; adaptor protein complex AP-3 beta-1 subunit; adaptor protein complex AP-3 subunit beta-1; adaptor related protein complex 3 beta 1 subunit; adaptor related protein complex 3 subunit beta 1; adaptor related protein complex 3, beta 1 subunit; adaptor-related protein complex 3 subunit beta-1; adaptor-related protein complex 3, beta 1 subunit; adaptor-related protein complex AP3 beta 1 subunit; adaptor-related protein complex AP-3, beta 1 subunit; ADTB3; ADTB3A; ap3; AP-3; AP-3 complex beta-3A subunit; AP-3 complex subunit beta-1; AP-3 complex subunit beta-1-like protein; AP3B1; AU015684; beta3A; Beta-3A-adaptin; beta3A-adaptin; C78395; Clathrin assembly protein complex 3 beta-1 large chain; HPS; Hps2; PE; pearl; recombination induced mutation 2; rim2; Unknown (protein for MGC:128992) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11774, 8546 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.49 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O00203, Q9Z1T1 | |
Ap3b1 | |
Recombinant fragment corresponding to a region within amino acids 33 and 281 of Human HPS2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction