Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP3B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558524
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558524 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558524 Supplier Invitrogen™ Supplier No. PA558524
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VISVSTPAFV PTKTHVLLHR MSGKGLAAHY FFPRQPCIFG DKMVSIQITL NNTTDRKIEN IHIGEKKLPI GMKMHVFNPI DSLEPEGS Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 80%.

AP3B1 is a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. Structurally, AP3B1 is part of the heterotetrameric AP-3 protein complex which interacts with the scaffolding protein clathrin. Mutations in AP3B1 are associated with Hermansky-Pudlak syndrome type 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP3B1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ap3b1
Gene Accession No. O00203
Gene Alias adapter-related protein complex 3 subunit beta-1; adaptor protein complex AP-3 beta-1 subunit; adaptor protein complex AP-3 subunit beta-1; adaptor related protein complex 3 beta 1 subunit; adaptor related protein complex 3 subunit beta 1; adaptor related protein complex 3, beta 1 subunit; adaptor-related protein complex 3 subunit beta-1; adaptor-related protein complex 3, beta 1 subunit; adaptor-related protein complex AP3 beta 1 subunit; adaptor-related protein complex AP-3, beta 1 subunit; ADTB3; ADTB3A; ap3; AP-3; AP-3 complex beta-3A subunit; AP-3 complex subunit beta-1; AP-3 complex subunit beta-1-like protein; AP3B1; AU015684; beta3A; Beta-3A-adaptin; beta3A-adaptin; C78395; Clathrin assembly protein complex 3 beta-1 large chain; HPS; Hps2; PE; pearl; recombination induced mutation 2; rim2; Unknown (protein for MGC:128992)
Gene Symbols Ap3b1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AP3B1. Recombinant protein control fragment (Product #RP-96853).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8546
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.