Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AP3B1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576207
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
AP3B1 is a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. Structurally, AP3B1 is part of the heterotetrameric AP-3 protein complex which interacts with the scaffolding protein clathrin. Mutations in AP3B1 are associated with Hermansky-Pudlak syndrome type 2.
Specifications
AP3B1 | |
Polyclonal | |
Unconjugated | |
Ap3b1 | |
adapter-related protein complex 3 subunit beta-1; adaptor protein complex AP-3 beta-1 subunit; adaptor protein complex AP-3 subunit beta-1; adaptor related protein complex 3 beta 1 subunit; adaptor related protein complex 3 subunit beta 1; adaptor related protein complex 3, beta 1 subunit; adaptor-related protein complex 3 subunit beta-1; adaptor-related protein complex 3, beta 1 subunit; adaptor-related protein complex AP3 beta 1 subunit; adaptor-related protein complex AP-3, beta 1 subunit; ADTB3; ADTB3A; ap3; AP-3; AP-3 complex beta-3A subunit; AP-3 complex subunit beta-1; AP-3 complex subunit beta-1-like protein; AP3B1; AU015684; beta3A; Beta-3A-adaptin; beta3A-adaptin; C78395; Clathrin assembly protein complex 3 beta-1 large chain; HPS; Hps2; PE; pearl; recombination induced mutation 2; rim2; Unknown (protein for MGC:128992) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11774, 8546 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O00203, Q9Z1T1 | |
Ap3b1 | |
Recombinant protein of human AP3B1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction