Learn More
AP3B1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305349AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
AP3B1 is a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. Structurally, AP3B1 is part of the heterotetrameric AP-3 protein complex which interacts with the scaffolding protein clathrin. Mutations in AP3B1 are associated with Hermansky-Pudlak syndrome type 2.Specifications
AP3B1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O00203 | |
AP3B1 | |
Between 725 and 775 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
AP3B1 | |
ADTB3, ADTB3A, HPS, HPS2, PE, AP-3 complex beta-3A subunit, AP-3 complex subunit beta-1, adapter-related protein complex 3 subunit beta-1, adaptor protein complex AP-3 subunit beta-1, beta-3A-adaptin, beta3A-adaptin, clathrin assembly protein complex 3 beta-1 large chain | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8546 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.