Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP3B1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501570193
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-0193 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0193 Supplier Bethyl Laboratories, Inc Supplier No. A305349A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

AP3B1 is a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. Structurally, AP3B1 is part of the heterotetrameric AP-3 protein complex which interacts with the scaffolding protein clathrin. Mutations in AP3B1 are associated with Hermansky-Pudlak syndrome type 2.

Specifications

Antigen AP3B1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene AP3B1
Gene Accession No. O00203
Gene Alias ADTB3, ADTB3A, HPS, HPS2, PE, AP-3 complex beta-3A subunit, AP-3 complex subunit beta-1, adapter-related protein complex 3 subunit beta-1, adaptor protein complex AP-3 subunit beta-1, beta-3A-adaptin, beta3A-adaptin, clathrin assembly protein complex 3 beta-1 large chain
Gene Symbols AP3B1
Host Species Rabbit
Immunogen Between 725 and 775
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8546
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.