Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP3B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111115
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111115 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111115 Supplier Invitrogen™ Supplier No. PA5111115
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPEWTKCSNR EKRKEKEKPF YSDSEGESGP TESADSDPES ESESDSKSSS ESGSGESSSE SDNEDQ Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 92%.

The AP3 complex is a heterotetramer composed of two large adaptins (AP3D1, AP3B1 or AP3B2), a medium adaptin (AP3M1 or AP3M2) and a small adaptin (APS1 or AP3S2). The complex is associated with the Golgi region as well as more peripheral structures. It facilitates the budding of vesicles from the Golgi membrane and may be directly involved in trafficking to lysosomes. Unlike the homologous AP3B1, AP3B2-containing AP3 complexes are preferentially targeted to neuronal processes, and mice deficient in AP3B2 displayed compromised synaptic zinc stores.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP3B2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AP3B2
Gene Accession No. Q13367
Gene Alias [b]-NAP; adapter-related protein complex 3 subunit beta-2; adaptor protein complex AP-3 beta-2 subunit; adaptor protein complex AP-3 subunit beta-2; adaptor related protein complex 3 beta 2 subunit; adaptor related protein complex 3, beta 2 subunit; adaptor-related protein complex 3 subunit beta-2; adaptor-related protein complex 3, beta 2 subunit; adaptor-related protein complex AP-3, beta 2 subunit; AI549966; ap3; AP-3 complex beta-2 subunit; AP-3 complex subunit beta-2; Ap3b2; AU042881; beta3B; beta-3B-adaptin; beta3B-adaptin; beta-NAP; Clathrin a; clathrin assembly protein complex 3 beta-2 large chain; NAPTB; Neuronal adaptin-like protein, beta-subunit; neuron-specific vesicle coat protein beta-NAP
Gene Symbols AP3B2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human AP3B2. Recombinant protein control fragment (Product #RP-98025).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8120
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.