Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AP3D1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100384
Description
Antibody detects endogenous levels of total AP3D1.
AP3D1 is a subunit of the AP3 adaptor-like complex, which is not associated with clathrin. The AP3D1 subunit is implicated in intracellular biogenesis and trafficking of pigment granules and possibly platelet dense granules and neurotransmitter vesicles.
Specifications
AP3D1 | |
Polyclonal | |
Unconjugated | |
Ap3d1 | |
AA407035; adapter-related protein complex 3 subunit delta-1; adaptin, delta; adaptor related protein complex 3 delta 1 subunit; adaptor related protein complex 3 subunit delta 1; adaptor related protein complex 3, delta 1 subunit; adaptor-related protein complex 3 subunit delta-1; adaptor-related protein complex 3, delta 1 subunit; adaptor-related protein complex 3, delta subunit; adaptor-related protein complex AP-3, delta subunit; ADTD; ap3; AP-3 complex delta subunit, partial CDS; AP-3 complex subunit delta; AP-3 complex subunit delta-1; Ap3d; Ap3d1; Bolvr; delta adaptin; delta-adaptin; delta-adaptin, partial CDS; hBLVR; mBLVR1; mh; mocha; PRO0039; subunit of putative vesicle coat adaptor complex AP-3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
11776, 314633, 8943 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O14617, O54774 | |
Ap3d1 | |
A synthesized peptide derived from human AP3D1(Accession O14617), corresponding to amino acid residues E766-L816. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction