Learn More
Invitrogen™ AP3M1 Polyclonal Antibody

Description
A suggested positive control is human brain tissue lysate. PA5-21162 can be used with blocking peptide PEP-1276.
Specifications
Specifications
| Antigen | AP3M1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Ap3m1 |
| Gene Accession No. | P53676, Q9JKC8, Q9Y2T2 |
| Gene Alias | 1200013D09Rik; adapter-related protein complex 3 mu-1 subunit; adapter-related protein complex 3 subunit mu-1; adaptor protein complex 3 Mu3A; adaptor related protein complex 3 mu 1 subunit; adaptor related protein complex 3 subunit mu 1; adaptor-related protein complex 3 subunit mu-1; adaptor-related protein complex 3, mu 1 subunit; adaptor-related protein complex AP-3 mu 1 subunit; adaptor-related protein complex AP-3, mu 1 subunit; ap3; AP-3 adapter complex mu3A subunit; AP-3 adaptor complex mu3A subunit; AP-3 complex subunit mu-1; AP-3 complex subunit mu-1-like protein; Ap3m1; C78982; clathrin adaptor complex AP3, mu-3A subunit; clathrin adaptor protein mu3A; clathrin assembly protein assembly protein complex 1 medium chain homolog 1; clathrin assembly protein assembly protein complex 3 mu-1 medium chain; clathrin coat assembly protein AP47 homolog 1; clathrin coat-associated protein AP47 homolog 1; clathrin-associated adaptor protein homolog (p47A); golgi adaptor AP-1 47 kDa protein homolog 1; HA1 47 kDa subunit homolog 1; hypothetical protein; Mu3A-adaptin; mu-adaptin 3A; P47a; R75378; RCJMB04_1h22; Unknown (protein for MGC:137644) |
| Gene Symbols | Ap3m1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.