Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AP3M1 Rabbit anti-Human, Clone: S08-8D2, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | AP3M1 |
---|---|
Clone | S08-8D2 |
Dilution | Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
AP3M1 Monoclonal antibody specifically detects AP3M1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
AP3M1 | |
Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunohistochemistry-Paraffin 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
adaptor-related protein complex 3, mu 1 subunit, AP-3 adapter complex mu3A subunit, MGC22164, mu-3A subunit, mu3A-adaptin | |
A synthetic peptide of human AP3M1 (Uniprot#: Q9Y2T2) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S08-8D2 | |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
26985 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title