Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AP3M2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543790
Description
Peptide sequence: VVNTITGSTN VGDQLPTGQL SVVPWRRTGV KYTNNEAYFD VIEEIDAIID Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
This gene encodes a subunit of the heterotetrameric adaptor-related protein complex 3 (AP-3), which belongs to the adaptor complexes medium subunits family. The AP-3 complex plays a role in protein trafficking to lysosomes and specialized organelles. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
AP3M2 | |
Polyclonal | |
Unconjugated | |
Ap3m2 | |
5830445E16Rik; adapter-related protein complex 3 mu-2 subunit; adapter-related protein complex 3 subunit mu-2; adaptor related protein complex 3 mu 2 subunit; adaptor related protein complex 3 subunit mu 2; adaptor related protein complex 3, mu 2 subunit; adaptor-related protein complex 3 subunit mu-2; adaptor-related protein complex 3, mu 2 subunit; AP-3 complex subunit mu-2; AP-3B; Ap3m2; AP47B; cla; CLA20; clathrin assembly protein assembly protein complex 1 medium chain homolog 2; clathrin assembly protein assembly protein complex 3 mu-2 medium chain; Clathrin coat assembly protein AP47 homolog 2; clathrin coat-associated protein AP47 homolog 2; clathrin-associated protein AP47 homolog 2; fc15g12; Golgi adaptor AP-1 47 kDa protein homolog 2; HA1 47 kDa subunit homolog 2; HA1 47kDA subunit homolog 2; m3B; Mu3B-adaptin; P47B; wu:fc15g12; zgc:86670 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10947 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P53677 | |
Ap3m2 | |
Synthetic peptide directed towards the middle region of human AP3M2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction