Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP3S1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521161

Catalog No. PIPA521161


Only null left
Add to Cart

Description

Description

A suggested positive control is mouse kidney tissue lysate. PA5-21161 can be used with blocking peptide PEP-1275.

Part of the AP-3 complex, an adapter-related complex which is not clathrin-associated. The complex is associated with the Golgi region as well as more peripheral structures. It facilitates the budding of vesicles from the Golgi membrane and may be directly involved in trafficking to lysosomes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AP3S1
Polyclonal
Unconjugated
AP3S1
adapter-related protein complex 3 subunit sigma-1; adaptor related protein complex 3 sigma 1 subunit; adaptor related protein complex 3, sigma 1 subunit; adaptor-related protein complex 3 subunit sigma-1; adaptor-related protein complex 3, sigma 1 subunit; AP-3 complex subunit sigma-1; AP-3 complex subunit sigma-3A; AP3S1; CLAPS3; clathrin adaptor complex AP3, sigma-3A subunit; clathrin-associated/assembly/adapter protein, small 3; Clathrin-associated/assembly/adaptor protein, small 3; clathrin-associated/assembly/adaptor protein, small 3 (22kD); Sigma3A; Sigma-3A-adaptin; Sigma3A-adaptin; Sigma-adaptin 3a
Rabbit
Antigen Affinity Chromatography
RUO
1176, 11777, 302290
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q92572, Q9DCR2
AP3S1
A 19 amino acid peptide near the center of human AP3S1.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.