Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP3S2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597466
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597466 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597466 Supplier Invitrogen™ Supplier No. PA597466
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AP3S2 Polyclonal Antibody for Western Blot, ELISA

AP3S2 (Adaptor Related Protein Complex 3 Subunit Sigma 2) is a Protein Coding gene. Among its related pathways are Lysosome. Gene Ontology (GO) annotations related to this gene include protein transporter activity. An important paralog of this gene is AP3S1. Part of the AP-3 complex, an adaptor-related complex which is not clathrin-associated. The complex is associated with the Golgi region as well as more peripheral structures. It facilitates the budding of vesicles from the Golgi membrane and may be directly involved in trafficking to lysosomes. In concert with the BLOC-1 complex, AP-3 is required to target cargos into vesicles assembled at cell bodies for delivery into neurites and nerve terminals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP3S2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene AP3S2
Gene Accession No. P59780
Gene Alias [s]3B; adapter-related protein complex 3 sigma-2 subunit; adapter-related protein complex 3 subunit sigma-2; adaptor complex sigma3B; adaptor related protein complex 3 sigma 2 subunit; adaptor related protein complex 3, sigma 2 subunit; adaptor-related protein complex 3 subunit sigma-2; adaptor-related protein complex 3, sigma 2 subunit; adaptor-related protein complex AP-3, sigma 2 subunit; AP-3 complex sigma-3B subunit; AP-3 complex subunit sigma-2; AP-3 complex subunit sigma-3B; AP3S2; AP3S3; clathrin-associated/assembly/adaptor protein, small 4, 22-kD; FLJ35955; sigma 3B; sigma3b; sigma-3B-adaptin; sigma3B-adaptin; Sigma-adaptin 3b
Gene Symbols AP3S2
Host Species Rabbit
Immunogen Recombinant Human AP-3 complex subunit sigma-2 protein (1-193AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10239
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.