Learn More
AP3S2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
Specifications
Specifications
| Antigen | AP3S2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.15 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | AP3S2 |
| Gene Accession No. | P59780, Q8BSZ2 |
| Gene Alias | AP 3 complex subunit sigma 2, AP 3 complex subunit sigma 3B, AP3S2, AP3S3, Sigma 3B adaptin, Sigma adaptin 3b, sigma3b, Sigma3B adaptin |
| Gene Symbols | Ap3s2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.