Learn More
AP4 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302761AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Transcription factors of the basic helix-loop-helix-zipper(bHLH-ZIP) family contain a basic domain, which is used for DNAbinding, and HLH and ZIP domains, which are used foroligomerization. Transcription factor AP4 activates both viral andcellular genes by binding to the symmetrical DNA sequence CAGCTG.Specifications
| AP4 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q01664 | |
| TFAP4 | |
| Between 288 and 338 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| TFAP4 | |
| AP-4, bHLHc41, activating enhancer-binding protein 4, class C basic helix-loop-helix protein 41, transcription factor AP-4, transcription factor AP-4 (activating enhancer-binding protein 4) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7023 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.