Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AP4B1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548836
Description
Predicted to react with mouse based on sequence homology.
Subunit of novel type of clathrin- or non-clathrin- associated protein coat involved in targeting proteins from the trans-Golgi network (TGN) to the endosomal-lysosomal system.
Specifications
AP4B1 | |
Polyclonal | |
Unconjugated | |
AP4B1 | |
adaptor related protein complex 4 beta 1 subunit; adaptor related protein complex 4, beta 1 subunit; Adaptor-related protein complex 4 subunit beta-1; adaptor-related protein complex 4, beta 1 subunit; AP-4 adaptor complex subunit beta; AP-4 complex subunit beta-1; AP4B1; beta 4 subunit of AP-4; Beta subunit of AP-4; BETA-4; Beta4-adaptin; CPSQ5; spastic paraplegia 47; SPG47 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10717 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mg | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9Y6B7 | |
AP4B1 | |
KLH conjugated synthetic peptide between 492-520 amino acids from the Central region of human AP4B1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction