Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP4B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113563
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113563 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113563 Supplier Invitrogen™ Supplier No. PA5113563
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AP4B1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Subunit of novel type of clathrin- or non-clathrin- associated protein coat involved in targeting proteins from the trans-Golgi network (TGN) to the endosomal-lysosomal system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP4B1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene AP4B1
Gene Accession No. Q9Y6B7
Gene Alias adaptor related protein complex 4 beta 1 subunit; adaptor related protein complex 4, beta 1 subunit; Adaptor-related protein complex 4 subunit beta-1; adaptor-related protein complex 4, beta 1 subunit; AP-4 adaptor complex subunit beta; AP-4 complex subunit beta-1; AP4B1; beta 4 subunit of AP-4; Beta subunit of AP-4; BETA-4; Beta4-adaptin; CPSQ5; spastic paraplegia 47; SPG47
Gene Symbols AP4B1
Host Species Rabbit
Immunogen Recombinant Human AP-4 complex subunit beta-1 protein (201-500AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10717
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.