Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP4E1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114335
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114335 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114335 Supplier Invitrogen™ Supplier No. PA5114335
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody specifically recognizes AP4E1 in Arabidopsis thaliana.

This gene encodes a member of the adaptor complexes large subunit protein family. These proteins are components of the heterotetrameric adaptor protein complexes, which play important roles in the secretory and endocytic pathways by mediating vesicle formation and sorting of integral membrane proteins. The encoded protein is a large subunit of adaptor protein complex-4, which is associated with both clathrin- and nonclathrin-coated vesicles. Disruption of this gene may be associated with cerebral palsy. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP4E1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Alias adaptor related protein complex 4 epsilon 1 subunit; adaptor related protein complex 4, epsilon 1 subunit; adaptor-related protein complex 4 subunit epsilon-1; adaptor-related protein complex 4, epsilon 1 subunit; adaptor-related protein complex AP-4 epsilon; AP-4 adaptor complex subunit epsilon; AP-4 complex subunit epsilon-1; AP4E1; CPSQ4; Epsilon subunit of AP-4; Epsilon-adaptin; SPG51; STUT1
Host Species Rabbit
Immunogen Synthetic peptide within thaliana AP-4 complex subunit epsilon aa 889-938.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.