Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AP4E1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119948
This item is not returnable.
View return policy
Description
Positive Control: A. thaliana tissue. Subcellular Location: Trans-Golgi network, coated pit.
This gene encodes a member of the adaptor complexes large subunit protein family. These proteins are components of the heterotetrameric adaptor protein complexes, which play important roles in the secretory and endocytic pathways by mediating vesicle formation and sorting of integral membrane proteins. The encoded protein is a large subunit of adaptor protein complex-4, which is associated with both clathrin- and nonclathrin-coated vesicles. Disruption of this gene may be associated with cerebral palsy. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| AP4E1 | |
| Polyclonal | |
| Unconjugated | |
| adaptor related protein complex 4 epsilon 1 subunit; adaptor related protein complex 4, epsilon 1 subunit; adaptor-related protein complex 4 subunit epsilon-1; adaptor-related protein complex 4, epsilon 1 subunit; adaptor-related protein complex AP-4 epsilon; AP-4 adaptor complex subunit epsilon; AP-4 complex subunit epsilon-1; AP4E1; CPSQ4; Epsilon subunit of AP-4; Epsilon-adaptin; SPG51; STUT1 | |
| Synthetic peptide within ARATH AP-4 complex subunit epsilon aa 880-938/938. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| Plant | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction