Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP4M1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119950
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119950 Supplier Invitrogen™ Supplier No. PA5119950
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: A. thaliana tissue. Subcellular Location: Trans-Golgi network, coated pit.

This gene encodes a subunit of the heterotetrameric AP-4 complex. The encoded protein belongs to the adaptor complexes medium subunits family. This AP-4 complex is involved in the recognition and sorting of cargo proteins with tyrosine-based motifs from the trans-Golgi network to the endosomal-lysosomal system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP4M1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Alias 4930443L05Rik; adapter-related protein complex 4 mu-1 subunit; adapter-related protein complex 4 subunit mu-1; adaptor related protein complex 4 mu 1 subunit; adaptor related protein complex 4, mu 1 subunit; adaptor-related protein complex 4 subunit mu-1; adaptor-related protein complex 4, mu 1 subunit; adaptor-related protein complex AP-4 mu4 subunit; adaptor-related protein complex AP-4, mu 1; AP-4 adapter complex mu subunit; AP-4 adaptor complex mu subunit; AP-4 complex subunit mu-1; Ap4m1; ap4m1 {ECO:0000312; Ap4m4; CPSQ3; Mu subunit of AP-4; mu4; MU-4; mu4-adaptin; Mu-adaptin-related protein 2; mu-adaptin-related protein-2; MUARP2; mu-ARP2; RGD:1310233}; SPG50
Host Species Rabbit
Immunogen Synthetic peptide within A. thaliana AP-4 complex subunit mu aa 380-430.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.