Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AP4M1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 116531AP150UL
Description
This gene encodes a subunit of the heterotetrameric AP-4 complex. The encoded protein belongs to the adaptor complexes medium subunits family. This AP-4 complex is involved in the recognition and sorting of cargo proteins with tyrosine-based motifs from the trans-Golgi network to the endosomal-lysosomal system.Specifications
| AP4M1 | |
| Polyclonal | |
| Unconjugated | |
| AP4M1 | |
| AP 4 complex subunit mu 1, AP4M1, MU 4, Mu adaptin related protein 2, MU ARP2, Mu subunit of AP 4, mu4, Mu4 adaptin, MUARP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11781, 304344, 9179 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O00189, Q2PWT8, Q9JKC7 | |
| AP4M1 | |
| AP4M1 Fusion Protein Ag2222 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction